accessibility menu, dialog, popup

UserName

CH3OOOCH3

CAS RN 1131-52-8

IUPAC Name: 3-ethoxy-4-methoxybenzaldehyde
Synonyms: 3-ethoxy-4-methoxybenzaldehyde
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: VAMZHXWLGRQSJS-UHFFFAOYSA-N
SMILES: CCOC1=CC(C=O)=CC=C1OC
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)