missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-4-methoxybenzaldehyde 98.0+%, TCI America™

Catalog No. p-7129176
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08355G 5 g
E083525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08355G Supplier TCI America Supplier No. E08355G
Only null left

Chemical Identifiers

CAS 1131-52-8
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00010128
InChI Key VAMZHXWLGRQSJS-UHFFFAOYSA-N
Synonym 3-ethoxy-4-methoxy-benzaldehyde, benzaldehyde, 3-ethoxy-4-methoxy, pubchem13135, 3-ethoxy-p-anisaldehyde, acmc-1ca82, 3-ethoxy-4methoxybenzaldehyde, 3ethoxy-4-methoxybenzaldehyde, ksc183c6j, 3-ethoxy-4-methoxybenzaldehyd, 4-methoxy-3-ethoxybenzaldehyde
PubChem CID 244728
IUPAC Name 3-ethoxy-4-methoxybenzaldehyde
SMILES CCOC1=C(C=CC(=C1)C=O)OC

Specifications

Melting Point 50°C
Color Yellow
Boiling Point 155°C
Quantity 5 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Ethoxy-4-methoxybenzaldehyde