missing translation for 'onlineSavingsMsg'
Learn More

3-(Dimethylsulfamoyl)benzeneboronic acid, 98%

Catalog No. AAH52838MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52838MD 250 mg
AAH5283803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52838MD Supplier Thermo Scientific Chemicals Supplier No. H52838MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 871329-59-8 | C8H12BNO4S | 229.057 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 871329-59-8
Molecular Formula C8H12BNO4S
Molecular Weight (g/mol) 229.057
MDL Number MFCD07363747
InChI Key SGERXKCDWJPIOS-UHFFFAOYSA-N
Synonym 3-n,n-dimethylsulfamoyl phenyl boronic acid, n,n-dimethyl 3-boronobenzenesulfonamide, 3-dimethylsulfamoyl phenyl boronic acid, 3-n,n-dimethylsulphonamido benzeneboronic acid, 3-dimethylsulfamoyl phenylboronic acid, 3-dimethylsulfamoyl benzeneboronic acid, 3-n,n-dimethylsulfonamidophenyl boronic acid, boronic acid, 3-dimethylamino sulfonyl phenyl, boronic acid,b-3-dimethylamino sulfonyl phenyl, acmc-209qg4
PubChem CID 44119634
IUPAC Name [3-(dimethylsulfamoyl)phenyl]boronic acid
SMILES B(C1=CC(=CC=C1)S(=O)(=O)N(C)C)(O)O

Specifications

Melting Point 88°C to 92°C
Quantity 250 mg
Formula Weight 229.06
Percent Purity 98%
Chemical Name or Material 3-(Dimethylsulfamoyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only