missing translation for 'onlineSavingsMsg'
Learn More

3-(Dimethylsilyloxy)-1,1,5,5-tetramethyl-3-phenyltrisiloxane 96.0+%, TCI America™

Catalog No. D376225ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D376225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D376225ML Supplier TCI America Supplier No. D376225ML
Only null left

Chemical Identifiers

CAS 18027-45-7
Molecular Formula C12H23O3Si4
Molecular Weight (g/mol) 327.65
MDL Number MFCD00053849
InChI Key YKSADNUOSVJOAS-UHFFFAOYSA-N
Synonym Tris(dimethylsilyloxy)phenylsilane
PubChem CID 6335366
IUPAC Name [bis[(dimethyl-$l^{3}-silanyl)oxy]-phenylsilyl]oxy-dimethylsilicon
SMILES C[Si](C)O[Si](O[Si](C)C)(O[Si](C)C)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 91°C
Quantity 25 mL
Formula Weight 330.68
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Dimethylsilyloxy)-1,1,5,5-tetramethyl-3-phenyltrisiloxane