missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylaminoacrolein, 90%

Catalog No. AC433320250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC433320250 25 g Glass bottle
AC433320050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC433320250 Supplier Thermo Scientific Chemicals Supplier No. 433320250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 927-63-9 | C5H9NO | 99.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 927-63-9
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00006999
InChI Key RRLMPLDPCKRASL-ONEGZZNKSA-N
Synonym 3-dimethylaminoacrolein, 3-dimethylaminoacrylaldehyde, 3-dimethylamino acrylaldehyde, 3-dimethylamino acrolein, 2-propenal, 3-dimethylamino, e-3-dimethylamino acrylaldehyde, e-3-dimethylamino prop-2-enal, 3-dimethylamino propenal, 2e-3-dimethylamino prop-2-enal, .beta.-dimethylamino acrolein
PubChem CID 638320
SMILES CN(C)\C=C\C=O

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Yellow to Brown
Density 0.7700g/mL
Boiling Point 270°C to 273°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Quantity 25 g
Specific Gravity 0.77
IUPAC Name (2E)-3-(dimethylamino)prop-2-enal
Formula Weight 99.13
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 3-Dimethylaminoacrolein
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-157-7

RUO – Research Use Only