missing translation for 'onlineSavingsMsg'
Learn More

3-(Dimethylamino)propyl Chloride Hydrochloride 98.0+%, TCI America™

Catalog No. D066325G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D066325G 25 g
D0663250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D066325G Supplier TCI America Supplier No. D066325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4-5-5407
Molecular Formula C5H13Cl2N
Molecular Weight (g/mol) 158.07
MDL Number MFCD00012521
InChI Key LJQNMDZRCXJETK-UHFFFAOYSA-N
Synonym 3-dimethylaminopropylchloride hydrochloride, 3-chloro-n,n-dimethylpropan-1-amine hydrochloride, 3-dimethylaminopropyl chloride hydrochloride, 3-dimethylamino propyl chloride hydrochloride, n,n-dimethyl-3-chloropropylamine hydrochloride, 3-chloropropyl dimethylamine hydrochloride, 3-chloro-n,n-dimethylpropylamine hydrochloride, dimethylaminopropyl chloride hydrochloride, ccris 7054, dimethylaminopropyl chloride, hydrochloride
PubChem CID 94308
IUPAC Name 3-chloro-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CN(C)CCCCl.Cl

Specifications

Melting Point 143°C
Color White
Quantity 25 g
Formula Weight 158.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Dimethylamino)propyl Chloride Hydrochloride
EINECSNumber (2)-0177&(9)-2009
RTECSNumber UI0209950
TSCA Yes