missing translation for 'onlineSavingsMsg'
Learn More

3-(Dimethylamino)-1,2-propanediol 98.0+%, TCI America™

Catalog No. D207225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D207225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D207225G Supplier TCI America Supplier No. D207225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 623-57-4
Molecular Formula C5H13NO2
Molecular Weight (g/mol) 119.164
MDL Number MFCD00004714
InChI Key QCMHUGYTOGXZIW-UHFFFAOYSA-N
Synonym N,N-Dimethyl-2,3-dihydroxypropylamine
PubChem CID 79078
IUPAC Name 3-(dimethylamino)propane-1,2-diol
SMILES CN(C)CC(CO)O

Specifications

Color Yellow
Boiling Point 217°C
Quantity 25 g
UN Number 2735
Formula Weight 119.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(Dimethylamino)-1,2-propanediol