missing translation for 'onlineSavingsMsg'
Learn More

3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one 98.0+%, TCI America™

Catalog No. D32625G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D32625G Supplier TCI America Supplier No. D32625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 165534-43-0
Molecular Formula C11H14N3O5P
Molecular Weight (g/mol) 299.223
InChI Key AJDPNPAGZMZOMN-UHFFFAOYSA-N
Synonym depbt, 3-diethoxyphosphoryloxy-1,2,3-benzotriazin-4 3h-one, 3-diethoxyphosphoryloxy-1,2,3-benzotrizin-4 3h-one, diethyl 4-oxobenzo d 1,2,3 triazin-3 4h-yl phosphate, diethyl 4-oxo-1,2,3-benzotriazin-3-yl phosphate, 3-diethoxyphosphoryloxy-3h-benzo d 1,2,3 triazin-4-one, 3-diethoxyphosphinyl oxy-1,2,3-benzotriazin-4 3h-one, 3-diethoxyphosphoryl oxy-1,2,3-benzotriazin-4 3h-one, 3-diethoxy-phosphoryloxy-3h-benzo d 1,2,3 triazin-4-one
PubChem CID 4293995
IUPAC Name diethyl (4-oxo-1,2,3-benzotriazin-3-yl) phosphate
SMILES CCOP(=O)(OCC)ON1C(=O)C2=CC=CC=C2N=N1

Specifications

Melting Point 73°C
Color White
Quantity 5 g
Formula Weight 299.22
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one