missing translation for 'onlineSavingsMsg'
Learn More

3-(Dibutylamino)propylamine 98.0+%, TCI America™

Catalog No. D1078500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1078500ML 500 mL
D107825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1078500ML Supplier TCI America Supplier No. D1078500ML

Chemical Identifiers

CAS 102-83-0
Molecular Formula C11H26N2
Molecular Weight (g/mol) 186.343
MDL Number MFCD00008219
InChI Key KYCGURZGBKFEQB-UHFFFAOYSA-N
Synonym 3-dibutylamino propylamine, n,n-dibutyl-1,3-propanediamine, n,n-dibutyltrimethylenediamine, 3-aminopropyl dibutylamine, 3-di-n-butylamino propylamine, n,n-dibutylaminopropylamine, n1,n1-dibutylpropane-1,3-diamine, 1,3-propanediamine, n,n-dibutyl, 3-aminopropyldibutylamine, 1,3-propanediamine, n1,n1-dibutyl
PubChem CID 1626
IUPAC Name N',N'-dibutylpropane-1,3-diamine
SMILES CCCCN(CCCC)CCCN

Specifications

Color Yellow
Boiling Point 125°C
Quantity 500 mL
UN Number 2922
Formula Weight 186.34
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(Dibutylamino)propylamine
EINECSNumber (2)-0145
RTECSNumber TX7175000
TSCA Yes