missing translation for 'onlineSavingsMsg'
Learn More

3-Deoxy-D-glucosone, 95%, Thermo Scientific Chemicals

Catalog No. AAJ66223LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66223LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66223LB0 Supplier Thermo Scientific Chemicals Supplier No. J66223LB0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Frequently used as a reference for the analysis and detection of glucose degradation products and glycating agents. Also used for cross-linking studies as it readily reacts with protein amino groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Frequently used as a reference for the analysis and detection of glucose degradation products and glycating agents
  • Also used for cross-linking studies as it readily reacts with protein amino groups

Chemical Identifiers

CAS 4084-27-9
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.141
MDL Number MFCD01723101
InChI Key ZGCHLOWZNKRZSN-NTSWFWBYSA-N
Synonym 3-deoxyglucosone, 3-deoxy-d-glucosone, 3-deoxy-d-erythro-hexos-2-ulose, d-3-deoxyglucosone, 3-deoxy-d-erythro-hexosulose, 3-deoxy-d-fructose, ccris 4272, 3-deoxy-2-ketoglucose, d-erythro-hexos-2-ulose, 3-deoxy, unii-exv5374vey
PubChem CID 114839
ChEBI CHEBI:60777
IUPAC Name (4S,5R)-4,5,6-trihydroxy-2-oxohexanal
SMILES C(C(C(CO)O)O)C(=O)C=O

Specifications

Color Yellow
Quantity 5 mg
Solubility Information Soluble in water,methanol or hot ethanol.
Formula Weight 162.14
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Deoxy-D-glucosone
RTECSNumber MQ3390000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only