missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanopyridine, 98%

Numéro de catalogue. p-7022202
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAA1485022 100 g
AAA1485036 500 g
AAA148500E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AAA1485022 Fournisseur Thermo Scientific Chemicals Code fournisseur A1485022
Il en reste null

CAS: 100-54-9 | C6H4N2 | 104.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 100-54-9
Molecular Formula C6H4N2
Molecular Weight (g/mol) 104.11
MDL Number MFCD00006372
InChI Key GZPHSAQLYPIAIN-UHFFFAOYSA-N
Synonym 3-cyanopyridine, nicotinonitrile, 3-pyridinecarbonitrile, nicotinic acid nitrile, 3-pyridinenitrile, 3-pyridylcarbonitrile, 3-azabenzonitrile, 3-cyjanopirydyna, nitryl kwasu nikotynowego, 3-cyjanopirydyna polish
PubChem CID 79
ChEBI CHEBI:86556
IUPAC Name pyridine-3-carbonitrile
SMILES N#CC1=CC=CN=C1

Spécifications

Melting Point 48°C to 51°C
Density 1.16
Boiling Point 206°C to 208°C
Flash Point 84°C (183°F)
Odor Pungent
Quantity 100 g
Beilstein 107711
Formula Weight 104.11
Percent Purity 98%
Chemical Name or Material 3-Cyanopyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 202-863-0
RTECSNumber QT3030000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only