missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanopyridine-4-boronic acid pinacol ester, 95%

Catalog No. AAH27814MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27814MD 250 mg
AAH2781403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27814MD Supplier Thermo Scientific Chemicals Supplier No. H27814MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 878194-92-4 | C12H15BN2O2 | 230.074 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 878194-92-4
Molecular Formula C12H15BN2O2
Molecular Weight (g/mol) 230.074
MDL Number MFCD09037481
InChI Key LUDBQHAQFBWGNI-UHFFFAOYSA-N
Synonym 3-cyanopyridine-4-boronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nicotinonitrile, 3-cyano-4-pyridineboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile, 3-cyanopyridin-4-ylboronic acid pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile, 3-cyano-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 3-pyridinecarbonitrile, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3-pyridinecarbonitrile, 3-cyanopyridine-4-boronicacidpinacolester
PubChem CID 44755176
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbonitrile
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=NC=C2)C#N

Specifications

Melting Point 97°C to 99°C
Quantity 250 mg
Formula Weight 230.07
Percent Purity 95%
Chemical Name or Material 3-Cyanopyridine-4-boronic acid pinacol ester
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only