missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-4-methylpyridine, 97%

Catalog No. AAL1976914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1976914 25 g
AAL1976903 1 g
AAL1976906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1976914 Supplier Thermo Scientific Chemicals Supplier No. L1976914
Only null left

CAS: 5444-01-9 | C7H6N2 | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5444-01-9
MDL Number MFCD00234272

Specifications

Melting Point 41°C to 44°C
Boiling Point 108°C to 111°C (20 mmHg)
Odor Characteristic
Molecular Formula C9H10O4
Quantity 25 g
Beilstein 112016
Synonym methyl 4-methoxysalicylate, 2-hydroxy-4-methoxy-benzoic acid methyl ester, methyl-4-methoxysalicylate, benzoic acid, 2-hydroxy-4-methoxy-, methyl ester, 2-hydroxy-4-methoxybenzoic acid methyl ester, 4-methoxysalicylic acid methyl ester, acmc-2097hp, methyl 2-hydroxy-p-anisate, ksc494a7p, 2-methoxycarbonyl-5-methoxyphenol
InChI Key ZICRWXFGZCVTBZ-UHFFFAOYSA-N
SMILES COC1=CC(=C(C=C1)C(=O)OC)O
IUPAC Name methyl 2-hydroxy-4-methoxybenzoate
Molecular Weight (g/mol) 182.18
PubChem CID 79523
Formula Weight 118.14
Percent Purity 97%
Chemical Name or Material 3-Cyano-4-methylpyridine
Show More Show Less
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only