missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-2-fluoropyridine 98.0+%, TCI America™

Catalog No. C25385G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25385G 5 g
C253825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25385G Supplier TCI America Supplier No. C25385G

Chemical Identifiers

CAS 3939-13-7
Molecular Formula C6H3FN2
Molecular Weight (g/mol) 122.102
MDL Number MFCD03095082
InChI Key USIDQCCXMGJOJM-UHFFFAOYSA-N
Synonym 3-cyano-2-fluoropyridine, 2-fluoronicotinonitrile, 2-fluoro-3-cyanopyridine, 3-pyridinecarbonitrile, 2-fluoro, 2-fluoro-nicotinonitrile, 2-fluoro-3-pyridinecarbonitrile, pubchem1108, 2-fluoronicotin-onitrile, acmc-1ckgi, 2-fluoro-3-nitrylpyridine
PubChem CID 2783254
IUPAC Name 2-fluoropyridine-3-carbonitrile
SMILES C1=CC(=C(N=C1)F)C#N

Specifications

Color Green-White
Quantity 5 g
Formula Weight 122.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Cyano-2-fluoropyridine