missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-2,4-difluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C3226200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3226200MG 200 mg
C32261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C3226200MG Supplier TCI America Supplier No. C3226200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 871940-31-7
Molecular Formula C7H4BF2NO2
Molecular Weight (g/mol) 182.921
MDL Number MFCD12026699
InChI Key VQEKFJVJHSVFTM-UHFFFAOYSA-N
Synonym 3-Cyano-2,4-difluorobenzeneboronic Acid
PubChem CID 44558118
IUPAC Name (3-cyano-2,4-difluorophenyl)boronic acid
SMILES B(C1=C(C(=C(C=C1)F)C#N)F)(O)O

Specifications

Melting Point 134°C
Color White
Quantity 200 mg
Formula Weight 182.92
Physical Form Crystalline Powder
Chemical Name or Material 3-Cyano-2,4-difluorophenylboronic Acid (contains varying amounts of Anhydride)