missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropyridine 95.0+%, TCI America™

Catalog No. C0280500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0280500G 500 g
C028025G 25 g
C0280100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0280500G Supplier TCI America Supplier No. C0280500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 626-60-8
Molecular Formula C5H4ClN
Molecular Weight (g/mol) 113.54
MDL Number MFCD00006375
InChI Key PWRBCZZQRRPXAB-UHFFFAOYSA-N
Synonym pyridine, 3-chloro, m-chloropyridine, unii-1m13huc1p4, ccris 1715, chloropyridine, b-chloropyridine, 3-chloro pyridine, 3-chloro-pyridine, 3-chloropyridine, 3-chloranylpyridine
PubChem CID 12287
IUPAC Name 3-chloropyridine
SMILES ClC1=CC=CN=C1

Specifications

Color Yellow
Boiling Point 148°C
Quantity 500 g
Formula Weight 113.54
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloropyridine
RTECSNumber US6125000
TSCA Yes