missing translation for 'onlineSavingsMsg'
Learn More

3-chlorophenylacetylene, 97%

Catalog No. AC433130050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC433130050 5 g Glass bottle
AC433130010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC433130050 Supplier Thermo Scientific Chemicals Supplier No. 433130050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 766-83-6 | C8H5Cl | 136.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 766-83-6
Molecular Formula C8H5Cl
Molecular Weight (g/mol) 136.58
MDL Number MFCD03839984
InChI Key GRBJPHPMYOUMJV-UHFFFAOYSA-N
Synonym 3-chlorophenylacetylene, 3-chlorophenyl acetylene, 3'-chlorophenyl acetylene, 3-chloro-1-ethynylbenzene, benzene, 1-chloro-3-ethynyl, m-chlorophenylacetylene, 1-chloro-3-ethynyl-benzene, 3'-chlorophenylacetylene, 3-chloro phenyl acetylene
PubChem CID 123086
IUPAC Name 1-chloro-3-ethynylbenzene
SMILES C#CC1=CC(=CC=C1)Cl

Specifications

Color Colorless to Yellow
CAS Min % 96.0
CAS Max % 100.0
Quantity 5 g
Boiling Point 178°C to 179°C
Flash Point 63°C
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Assay Percent Range 96% min. (GC)
Formula Weight 136.58
Percent Purity 97%
Chemical Name or Material 3-chlorophenylacetylene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only