missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3-Chlorophenyl)thiourea 98.0+%, TCI America™
Supplier: TCI America C31685G
Specifications
| (3-Chlorophenyl)thiourea | |
| 139°C | |
| C7H7ClN2S | |
| 5 g | |
| 1-3-chlorophenyl thiourea, 3-chlorophenyl thiourea, n-3-chlorophenyl thiourea, m-chlorophenyl thiourea, thiourea, 3-chlorophenyl, 1-m-chlorophenyl thiourea, urea, 1-m-chlorophenyl-2-thio, 1-3-chlorophenyl-2-thiourea | |
| C1=CC(=CC(=C1)Cl)NC(=S)N | |
| 186.657 | |
| 186.66 | |
| Crystalline Powder |
| 4947-89-1 | |
| White | |
| MFCD00022165 | |
| 2811 | |
| LSYZRUOXXOTVAV-UHFFFAOYSA-N | |
| (3-chlorophenyl)thiourea | |
| 2734794 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 4947-89-1 | |
| 186.657 | |
| LSYZRUOXXOTVAV-UHFFFAOYSA-N | |
| 2734794 | |
| C1=CC(=CC(=C1)Cl)NC(=S)N |
| C7H7ClN2S | |
| MFCD00022165 | |
| 1-3-chlorophenyl thiourea, 3-chlorophenyl thiourea, n-3-chlorophenyl thiourea, m-chlorophenyl thiourea, thiourea, 3-chlorophenyl, 1-m-chlorophenyl thiourea, urea, 1-m-chlorophenyl-2-thio, 1-3-chlorophenyl-2-thiourea | |
| (3-chlorophenyl)thiourea |
Safety and Handling
RTECSNumber : YS7000000
TSCA : No