missing translation for 'onlineSavingsMsg'
Learn More

(3-Chlorophenyl)thiourea 98.0+%, TCI America™

Catalog No. p-7125870
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31681G 1 g
C31685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31681G Supplier TCI America Supplier No. C31681G
Only null left

Chemical Identifiers

CAS 4947-89-1
Molecular Formula C7H7ClN2S
Molecular Weight (g/mol) 186.657
MDL Number MFCD00022165
InChI Key LSYZRUOXXOTVAV-UHFFFAOYSA-N
Synonym 1-3-chlorophenyl thiourea, 3-chlorophenyl thiourea, n-3-chlorophenyl thiourea, m-chlorophenyl thiourea, thiourea, 3-chlorophenyl, 1-m-chlorophenyl thiourea, urea, 1-m-chlorophenyl-2-thio, 1-3-chlorophenyl-2-thiourea
PubChem CID 2734794
IUPAC Name (3-chlorophenyl)thiourea
SMILES C1=CC(=CC(=C1)Cl)NC(=S)N

Specifications

Melting Point 139°C
Color White
Quantity 1 g
UN Number 2811
Formula Weight 186.66
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (3-Chlorophenyl)thiourea
RTECSNumber YS7000000
TSCA No