missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)-1-methyl-5-thien-2-yl-1H-pyrazole, ≥97%, Thermo Scientific™

Numéro de catalogue. p-3174975
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC47124CB 250mg Amber Glass Bottle
CC47124DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC47124CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC47124CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 876316-61-9
Formule moléculaire C9H9ClN2S
Poids moléculaire (g/mol) 212.70
Numéro MDL MFCD08435883
Clé InChI ODLZAEGCJDMPOT-UHFFFAOYSA-N
Synonyme 3-chloromethyl-1-methyl-5-thien-2-yl-1h-pyrazole, 3-chloromethyl-1-methyl-5-thiophen-2-yl pyrazole, 1h-pyrazole,3-chloromethyl-1-methyl-5-2-thienyl, 3-chloromethyl-1-methyl-5-thiophen-2-yl-1h-pyrazole, 2-3-chloromethyl-1-methylpyrazol-5-yl thiophene
PubChem CID 18525807
Nom de l’IUPAC 3-(chloromethyl)-1-methyl-5-thiophen-2-ylpyrazole
SOURIRES CN1N=C(CCl)C=C1C1=CC=CS1

Spécifications

Nom ou substance chimique 3-(Chloromethyl)-1-methyl-5-thien-2-yl-1H-pyrazole
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 250mg
Poids de formule 212.7
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.