missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Chloromethyl)-1,2,4-triazolin-5-one 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 199°C |
| Color | White |
| Quantity | 5 g |
| UN Number | 3261 |
| Formula Weight | 133.54 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-(Chloromethyl)-1,2,4-triazolin-5-one |
Safety and Handling
| TSCA | No |