missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzylideneacetone, 98%

Catalog No. AAA1033214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1033214 25 g
AAA1033206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1033214 Supplier Thermo Scientific Chemicals Supplier No. A1033214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20766-36-3 | C10H9ClO | 180.631 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20766-36-3
Molecular Formula C10H9ClO
Molecular Weight (g/mol) 180.631
MDL Number MFCD00052851
InChI Key VWEPXSRBBXPYSM-AATRIKPKSA-N
Synonym 3e-4-3-chlorophenyl but-3-en-2-one, e-4-3-chloro-phenyl-but-3-en-2-one, 4-3-chlorophenyl but-3-en-2-one, e-4-3-chlorophenyl-but-3-en-2-one, 3-chlorobenzylideneacetone, 3-chloro-benzalacetone, 3-chlorostyrylmethyl ketone, 3-chlorobenzylidene acetone, chembl73680, e-4-3-chlorophenyl but-3-en-2-one
PubChem CID 5373975
IUPAC Name (E)-4-(3-chlorophenyl)but-3-en-2-one
SMILES CC(=O)C=CC1=CC(=CC=C1)Cl

Specifications

Boiling Point 123°C to 125°C (1 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Formula Weight 180.64
Percent Purity 98%
Chemical Name or Material 3-Chlorobenzylideneacetone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only