Learn More
3-Chlorobenzonitrile, 99%
CAS: 766-84-7 | C7H4ClN | 137.57 g/mol
$109.10 - $1068.01
Chemical Identifiers
| CAS | 766-84-7 |
|---|---|
| Molecular Formula | C7H4ClN |
| Molecular Weight (g/mol) | 137.57 |
| MDL Number | MFCD00001798 |
| InChI Key | WBUOVKBZJOIOAE-UHFFFAOYSA-N |
| Synonym | m-chlorobenzonitrile, benzonitrile, 3-chloro, m-chloro benzonitrile, m-cyanochlorobenzene, benzonitrile, m-chloro, 3-chlorobenzenecarbonitrile, 5-chlorobenzonitrile, 3-chloro benzonitrile, 3-chloro-benzonitrile, pubchem3624 |
| PubChem CID | 13015 |
| IUPAC Name | 3-chlorobenzonitrile |
| SMILES | ClC1=CC=CC(=C1)C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1323709
|
Thermo Scientific Chemicals
A1323709 |
10 g |
Each for $109.10
|
|
|||||
|
AAA1323718
|
Thermo Scientific Chemicals
A1323718 |
50 g |
Each for $328.34
|
|
|||||
|
AAA1323730
|
Thermo Scientific Chemicals
A1323730 |
250 g |
Each for $1,068.01
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 766-84-7 | |
| 137.57 | |
| WBUOVKBZJOIOAE-UHFFFAOYSA-N | |
| 13015 | |
| ClC1=CC=CC(=C1)C#N |
| C7H4ClN | |
| MFCD00001798 | |
| m-chlorobenzonitrile, benzonitrile, 3-chloro, m-chloro benzonitrile, m-cyanochlorobenzene, benzonitrile, m-chloro, 3-chlorobenzenecarbonitrile, 5-chlorobenzonitrile, 3-chloro benzonitrile, 3-chloro-benzonitrile, pubchem3624 | |
| 3-chlorobenzonitrile |
Specifications
| 39°C to 42°C | |
| 1.14 | |
| 10 g | |
| C7H4ClN | |
| UN3439 | |
| m-chlorobenzonitrile, benzonitrile, 3-chloro, m-chloro benzonitrile, m-cyanochlorobenzene, benzonitrile, m-chloro, 3-chlorobenzenecarbonitrile, 5-chlorobenzonitrile, 3-chloro benzonitrile, 3-chloro-benzonitrile, pubchem3624 | |
| ClC1=CC=CC(=C1)C#N | |
| 137.57 | |
| 137.57 | |
| 3-Chlorobenzonitrile |
| 766-84-7 | |
| 94°C (10 mmHg) | |
| Almond-like | |
| MFCD00001798 | |
| 1906694 | |
| WBUOVKBZJOIOAE-UHFFFAOYSA-N | |
| 3-chlorobenzonitrile | |
| 13015 | |
| 99% |
Safety and Handling
GHS H Statement
H311-H302-H319
Toxic in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P363-P501c
H302+H312-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 212-172-6
RTECSNumber : DI2600000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only