missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzonitrile, 99%

Catalog No. aaa1323718
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
250 g
50 g
Catalog No. Quantity
AAA1323718 50 g
AAA1323709 10 g
AAA1323730 250 g
3 options

Catalog No. AAA1323718

Supplier: Thermo Scientific Chemicals A1323718

Only null left

CAS: 766-84-7 | C7H4ClN | 137.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 766-84-7
Molecular Formula C7H4ClN
Molecular Weight (g/mol) 137.57
MDL Number MFCD00001798
InChI Key WBUOVKBZJOIOAE-UHFFFAOYSA-N
Synonym m-chlorobenzonitrile, benzonitrile, 3-chloro, m-chloro benzonitrile, m-cyanochlorobenzene, benzonitrile, m-chloro, 3-chlorobenzenecarbonitrile, 5-chlorobenzonitrile, 3-chloro benzonitrile, 3-chloro-benzonitrile, pubchem3624
PubChem CID 13015
IUPAC Name 3-chlorobenzonitrile
SMILES ClC1=CC=CC(=C1)C#N

Specifications

Melting Point 39°C to 42°C
Density 1.14
Boiling Point 94°C (10 mmHg)
Quantity 50 g
Odor Almond-like
UN Number UN3439
Beilstein 1906694
Formula Weight 137.57
Percent Purity 99%
Chemical Name or Material 3-Chlorobenzonitrile
Hazard Category H302+H312-H319
Hazard Statement GHS H Statement
H311-H302-H319
Toxic in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 212-172-6
RTECSNumber DI2600000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only