missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzonitrile, 99%

CAS: 766-84-7 | C7H4ClN | 137.57 g/mol

Supplier:  Thermo Scientific Chemicals A1323730

 View more versions of this product

Catalog No. AAA1323730


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Chlorobenzonitrile
766-84-7
94°C (10 mmHg)
Almond-like
MFCD00001798
1906694
WBUOVKBZJOIOAE-UHFFFAOYSA-N
3-chlorobenzonitrile
13015
99%
39°C to 42°C
1.14
250 g
C7H4ClN
UN3439
m-chlorobenzonitrile, benzonitrile, 3-chloro, m-chloro benzonitrile, m-cyanochlorobenzene, benzonitrile, m-chloro, 3-chlorobenzenecarbonitrile, 5-chlorobenzonitrile, 3-chloro benzonitrile, 3-chloro-benzonitrile, pubchem3624
ClC1=CC=CC(=C1)C#N
137.57
137.57

Chemical Identifiers

766-84-7
137.57
WBUOVKBZJOIOAE-UHFFFAOYSA-N
13015
ClC1=CC=CC(=C1)C#N
C7H4ClN
MFCD00001798
m-chlorobenzonitrile, benzonitrile, 3-chloro, m-chloro benzonitrile, m-cyanochlorobenzene, benzonitrile, m-chloro, 3-chlorobenzenecarbonitrile, 5-chlorobenzonitrile, 3-chloro benzonitrile, 3-chloro-benzonitrile, pubchem3624
3-chlorobenzonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H319
Toxic in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P363-P501c

H302+H312-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 212-172-6

RTECSNumber : DI2600000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only