missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzoic Acid 99.0+%, TCI America™

Catalog No. C0132500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0132500G 500 g
C013225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0132500G Supplier TCI America Supplier No. C0132500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 535-80-8
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.565
MDL Number MFCD00002491
InChI Key LULAYUGMBFYYEX-UHFFFAOYSA-N
Synonym m-chlorobenzoic acid, benzoic acid, 3-chloro, benzoic acid, m-chloro, 3-chlorobenzoicacid, 3-chloro-benzoic acid, m-chlorobenzoate, acido m-clorobenzoico, unii-02uoj7064k, mcba, ccris 5992
PubChem CID 447
ChEBI CHEBI:49410
IUPAC Name 3-chlorobenzoic acid
SMILES C1=CC(=CC(=C1)Cl)C(=O)O

Specifications

Melting Point 157°C
Color White
Quantity 500 g
Formula Weight 156.57
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chlorobenzoic Acid
EINECSNumber (3)-0062&(3)-1424
RTECSNumber DG4975990
TSCA Yes