missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. C17915G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17915G 5 g
C17911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17915G Supplier TCI America Supplier No. C17915G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2888-06-4
Molecular Formula C6H4Cl2O2S
Molecular Weight (g/mol) 211.06
MDL Number MFCD00051697
InChI Key OINWZUJVEXUHCC-UHFFFAOYSA-N
Synonym 3-chlorobenzene-1-sulfonyl chloride, 3-chlorobenzenesulphonyl chloride, 3-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 3-chloro, m-chlorobenzenesulfonyl chloride, 3-chloro-benzenesulfonyl chloride, 3-chlorophenylsulfonyl chloride, pubchem5076, m-chlorbenzolsulfochlorid, acmc-1cj4b
PubChem CID 17909
IUPAC Name 3-chlorobenzenesulfonyl chloride
SMILES C1=CC(=CC(=C1)Cl)S(=O)(=O)Cl

Specifications

Color White-Yellow
Boiling Point 135°C
Quantity 5 g
UN Number 3261
Formula Weight 211.06
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Chlorobenzenesulfonyl Chloride