missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-N,N-dimethylaniline, 95%

Catalog No. AAL0856306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0856306 5 g
AAL0856314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0856306 Supplier Thermo Scientific Chemicals Supplier No. L0856306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6848-13-1 | C8H10ClN | 155.625 g/mol

It can be used to produce 3-(3-dimethylamino-phenyl)-acrylic acid butyl ester at the temperature of 140°C in the presence of reagent Na2CO3 and solvent N,N-dimethyl-acetamide with the reaction time of 20 hours.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used to produce 3-(3-dimethylamino-phenyl)-acrylic acid butyl ester at the temperature of 140°C in the presence of reagent Na2CO3 and solvent N,N-dimethyl-acetamide with the reaction time of 20 hours.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 6848-13-1
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.625
MDL Number MFCD00018456
InChI Key CHHCCYVOJBBCIY-UHFFFAOYSA-N
Synonym n,n-dimethyl-3-chloroaniline, 3-chloro-nn-dimethylaniline, benzenamine, 3-chloro-n,n-dimethyl, m-chloro-n,n-dimethylaniline, benzenamine,3-chloro-n,n-dimethyl, 3-chlorophenyl dimethylamine, pubchem3637, 3-chloranyl-n,n-dimethyl-aniline, 3-n,n-dimethylamino phenyl chloride
PubChem CID 23285
IUPAC Name 3-chloro-N,N-dimethylaniline
SMILES CN(C)C1=CC(=CC=C1)Cl

Specifications

Boiling Point 230°C to 232°C
Refractive Index 1.576
Quantity 5 g
UN Number UN2810
Beilstein 2206152
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 155.63
Percent Purity 95%
Chemical Name or Material 3-Chloro-N,N-dimethylaniline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 229-935-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only