missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-6-phenylpyridazine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 159°C |
| Quantity | 5 g |
| Formula Weight | 190.63 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Chloro-6-phenylpyridazine |
Safety and Handling
| TSCA | No |