missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-hydroxybenzonitrile 95.0+%, TCI America™

Catalog No. C27085G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27085G 5 g
C27081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27085G Supplier TCI America Supplier No. C27085G

Chemical Identifiers

CAS 473923-97-6
Molecular Formula C7H4ClNO
Molecular Weight (g/mol) 153.57
MDL Number MFCD11226540
InChI Key GHYUOOZZOMUNSY-UHFFFAOYSA-N
Synonym 3-Chloro-5-cyanophenol
PubChem CID 21949878
IUPAC Name 3-chloro-5-hydroxybenzonitrile
SMILES OC1=CC(Cl)=CC(=C1)C#N

Specifications

Melting Point 170°C
Color White-Yellow
Quantity 5 g
Formula Weight 153.57
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-5-hydroxybenzonitrile