missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-fluorobenzyl alcohol, 98+%

Catalog No. AAH3212206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3212206 5 g
AAH3212203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3212206 Supplier Thermo Scientific Chemicals Supplier No. H3212206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79944-64-2 | C7H6ClFO | 160.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79944-64-2
Molecular Formula C7H6ClFO
Molecular Weight (g/mol) 160.57
MDL Number MFCD03788554
InChI Key VJTJBAMDTCIMOB-UHFFFAOYSA-N
Synonym 3-chloro-5-fluorobenzyl alcohol, 3-chloro-5-fluorophenyl methanol, 3-chloro-5-fluoro-phenyl methanol, rarechem al bd 1247, benzenemethanol,3-chloro-5-fluoro, 3-chloro-5-fluoro phenyl methanol, 3-chloro-5-fluoro-phenyl-methanol, 3-chloro-5-fluorophenyl methan-1-ol
PubChem CID 2734835
IUPAC Name (3-chloro-5-fluorophenyl)methanol
SMILES OCC1=CC(F)=CC(Cl)=C1

Specifications

Refractive Index 1.531
Quantity 5 g
Formula Weight 160.58
Percent Purity ≥98%
Chemical Name or Material 3-Chloro-5-fluorobenzyl alcohol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only