missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-fluoroaniline 97.0+%, TCI America™

Catalog No. C30525G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30525G 5 g
C30521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30525G Supplier TCI America Supplier No. C30525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4863-91-6
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.561
MDL Number MFCD03407962
InChI Key LPIFAHAICWJMRR-UHFFFAOYSA-N
Synonym pubchem2953, acmc-209kdj, 3-chloro-5-fluorobenzenamine, 5-chloro-3-fluorophenylamine, ksc493m9f, 3-chloro-5-fluoro-phenylamine, buttpark 44\01-96
PubChem CID 2734838
IUPAC Name 3-chloro-5-fluoroaniline
SMILES C1=C(C=C(C=C1F)Cl)N

Specifications

Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 98°C
Formula Weight 145.56
Percent Purity ≥97.0% (GC,T)
Chemical Name or Material 3-Chloro-5-fluoroaniline