missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-hydroxybenzonitrile 98,0+%, TCI America™

Catalog No. C30831G
Change view
Click to view available options
Quantité:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
C30831G 1 g
C30835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30831G Supplier TCI America Supplier No. C30831G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2315-81-3
Formule moléculaire C7H4ClNO
Poids moléculaire (g/mol) 153.565
Numéro MDL MFCD01567246
Clé InChI CRYPJUOSZDQWJZ-UHFFFAOYSA-N
Synonyme benzonitrile, 3-chloro-4-hydroxy, 2-chloro-4-cyanophenol, 3-chloro-4-hydroxy-benzonitrile, 3-chloro-4-hydroxybenzenecarbonitrile, pubchem4726, 4-cyano-2-chlorophenol, buttpark 4357-94, acmc-1clj1, pharmabridge p-1867, buttpark 43\57-94
PubChem CID 2735739
Nom de l’IUPAC 3-chloro-4-hydroxybenzonitrile
SOURIRES C1=CC(=C(C=C1C#N)Cl)O

Specifications

Nom ou substance chimique 3-Chloro-4-hydroxybenzonitrile
Point de fusion 152 °C
Couleur Blanc-jaune
Quantité 1 g
Poids de formule 153.57
Pourcentage de pureté ≥98,0% (GC,T)
Forme physique Poudre cristalline