missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorothiobenzamide, 97%

Catalog No. AAL11132MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL11132MD 250 mg
AAL1113203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL11132MD Supplier Thermo Scientific Chemicals Supplier No. L11132MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 130560-97-3 | C7H5ClFNS | 189.632 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 130560-97-3
Molecular Formula C7H5ClFNS
Molecular Weight (g/mol) 189.632
MDL Number MFCD00042478
InChI Key RSGPCKCTKDCMRR-UHFFFAOYSA-N
Synonym 3-chloro-4-fluorothiobenzamide, 3-chloro-4-fluorobenzothioamide, 3-chloro-4-fluorobenzene-1-carbothioamide, benzenecarbothioamide,3-chloro-4-fluoro, amino 3-chloro-4-fluorophenyl methane-1-thione, acmc-1ccr0, 3-chloro-4-fluoro-thiobenzamide, #, 3-chloranyl-4-fluoranyl-benzenecarbothioamide, benzenecarbothioamide, 3-chloro-4-fluoro-9ci
PubChem CID 2779521
IUPAC Name 3-chloro-4-fluorobenzenecarbothioamide
SMILES C1=CC(=C(C=C1C(=S)N)Cl)F

Specifications

Melting Point 129°C to 130°C
Quantity 250 mg
UN Number UN2811
Beilstein 7757675
Formula Weight 189.64
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-fluorothiobenzamide
Hazard Category H302+H332
Hazard Statement GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only