missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4'-fluoropropiophenone, 97%

Catalog No. AAB2189122
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2189122 100 g
AAB2189106 5 g
AAB2189114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2189122 Supplier Thermo Scientific Chemicals Supplier No. B2189122
Only null left

CAS: 347-93-3 | C9H8ClFO | 186.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 347-93-3
Molecular Formula C9H8ClFO
Molecular Weight (g/mol) 186.61
MDL Number MFCD00000991
InChI Key AAHQPLJUSLMHHR-UHFFFAOYSA-N
Synonym 3-chloro-4'-fluoropropiophenone, 3-chloro-1-4-fluorophenyl propan-1-one, 3-chloro-1-4-fluorophenyl-1-propanone, 1-propanone, 3-chloro-1-4-fluorophenyl, p-fluoro-, a-chloropropiophenone, pubchem13250, acmc-1ctxg, ksc573a4j, 3-chloro-4'fluoropropiophenone, 3-chloro4'-fluoropropiophenone
PubChem CID 67672
IUPAC Name 3-chloro-1-(4-fluorophenyl)propan-1-one
SMILES C1=CC(=CC=C1C(=O)CCCl)F

Specifications

Melting Point 46°C to 49°C
Boiling Point 100°C to 103°C (3 mmHg)
Quantity 100 g
Beilstein 1866785
Formula Weight 186.61
Percent Purity 97%
Chemical Name or Material 3-Chloro-4'-fluoropropiophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-475-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only