missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-cyanobenzeneboronic acid, 96%

Catalog No. AAH61543MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61543MD 250 mg
AAH6154303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH61543MD Supplier Thermo Scientific Chemicals Supplier No. H61543MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1008415-02-8 | C7H5BClNO2 | 181.382 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1008415-02-8
Molecular Formula C7H5BClNO2
Molecular Weight (g/mol) 181.382
InChI Key QCJTUOIMDNJEHR-UHFFFAOYSA-N
Synonym 3-chloro-4-cyanophenyl boronic acid, boronic acid, b-3-chloro-4-cyanophenyl, acmc-2097sr, 3-chloro-4-cyanophenyl boronicacid
PubChem CID 46738932
IUPAC Name (3-chloro-4-cyanophenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)C#N)Cl)(O)O

Specifications

Quantity 250 mg
Formula Weight 164.93
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Chloro-4-cyanobenzeneboronic acid
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only