missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-pyridinecarboxylic Acid 98.0+%, TCI America™

Catalog No. C23425G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23425G 5 g
C23421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23425G Supplier TCI America Supplier No. C23425G

Chemical Identifiers

CAS 57266-69-0
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.553
MDL Number MFCD01646325
InChI Key XTMUXJBJCMRWPG-UHFFFAOYSA-N
Synonym 3-chloropicolinic acid, 3-chloro-2-pyridinecarboxylic acid, 3-chloro-2-picolinic acid, 2-pyridinecarboxylic acid, 3-chloro, 3-chloropyridine-2-carboxylicacid, 3-chloro-2-picolinicacid, 3-chloro-pyridine-2-carboxylic acid, chloropicolinic acid, pubchem12918, 3-chloro picolinic acid
PubChem CID 824632
IUPAC Name 3-chloropyridine-2-carboxylic acid
SMILES C1=CC(=C(N=C1)C(=O)O)Cl

Specifications

Melting Point 154°C
Color White-Yellow
Quantity 5 g
Formula Weight 157.55
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-2-pyridinecarboxylic Acid