missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-nitropyridine 98.0+%, TCI America™

Catalog No. p-7125157
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17651G 1 g
C17655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17651G Supplier TCI America Supplier No. C17651G

Chemical Identifiers

CAS 54231-32-2
Molecular Formula C5H3ClN2O2
Molecular Weight (g/mol) 158.541
MDL Number MFCD00490110
InChI Key PSGASDJUCYTRAD-UHFFFAOYSA-N
Synonym 3-chloro-2-nitro-pyridine, pyridine, 3-chloro-2-nitro, chloronitropyridine, chloro-nitropyridine, pubchem11172, acmc-209lej, 2-nitro-3-chloropyridine, 3-chloro-2-nitro pyridine, #
PubChem CID 554988
IUPAC Name 3-chloro-2-nitropyridine
SMILES C1=CC(=C(N=C1)[N+](=O)[O-])Cl

Specifications

Melting Point 91°C
Quantity 1 g
Formula Weight 158.54
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-2-nitropyridine