missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-methylbenzoic Acid 97.0+%, TCI America™

Catalog No. p-7125259
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20571G 1 g
C20575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20571G Supplier TCI America Supplier No. C20571G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7499-08-3
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00053312
InChI Key HXGHMCLCSPQMOR-UHFFFAOYSA-N
Synonym 3-chloro-o-toluic acid, 3-chloro-2-methyl-benzoic acid, benzoic acid, 3-chloro-2-methyl, o-toluic acid, 3-chloro, 3-chloro-2-methylbenzoicacid, 2-methyl-3-chlorobenzoic acid, pubchem8362, 2-carboxy-6-chlorotoluene, 3chloro-2-methylbenzoic acid, 3-chloro-2-methybenzoic acid
PubChem CID 82010
IUPAC Name 3-chloro-2-methylbenzoic acid
SMILES CC1=C(C=CC=C1Cl)C(=O)O

Specifications

Melting Point 164°C
Color White-Yellow
Quantity 1 g
Formula Weight 170.59
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-2-methylbenzoic Acid
RTECSNumber XU1645000
TSCA No