missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-2-chloromethyl-1-propene, 96%

Catalog No. AC277071000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC277071000 100 g Glass bottle
AC277070050 5 g Glass bottle
AC277070250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC277071000 Supplier Thermo Scientific Chemicals Supplier No. 277071000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1871-57-4 | C4H6Cl2 | 124.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1871-57-4
Molecular Formula C4H6Cl2
Molecular Weight (g/mol) 124.99
MDL Number MFCD00000952
InChI Key XJFZOSUFGSANIF-UHFFFAOYSA-N
Synonym 3-chloro-2-chloromethyl prop-1-ene, 3-chloro-2-chloromethyl-1-propene, 1,1-bis chloromethyl ethylene, methallyl dichloride, 1-propene, 3-chloro-2-chloromethyl, 2-chloromethyl-3-chloropropene, 1,3-dichloro-2-methylenepropane, 3-chloro-2-chloromethyl propene, 2-methylene-1,3-dichloropropane, 2-methylenepropane-1,3-dichloride
PubChem CID 15859
IUPAC Name 3-chloro-2-(chloromethyl)prop-1-ene
SMILES ClCC(=C)CCl

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 14°C
Color Colorless to Yellow
Density 1.0800g/mL
Boiling Point 138°C to 140°C
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4750 to 1.479
Linear Formula H2C=C(CH2Cl)2
Quantity 100 g
Beilstein 01, II, 181
Specific Gravity 1.08
Solubility Information Solubility in water: insoluble
Formula Weight 125
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-chloro-2-chloromethyl-1-propene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Toxic if swallowed.
Causes serious eye irritation.
Fatal if inhaled.
Very toxic to aquatic life.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 217-489-3

RUO – Research Use Only