missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-3-Carene 90,0+%, TCI America™
Spécifications
Spécifications
| Color | Jaune |
| Boiling Point | 170°C |
| Quantity | 25 mL |
| UN Number | 2319 |
| Formula Weight | 136.24 |
| Percent Purity | ≥90.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | (+)-3-Carene |
Sécurité et manipulation
| EINECSNumber | (4)-0589 |
| RTECSNumber | FH8400000 |
| TSCA | Yes |