missing translation for 'onlineSavingsMsg'
Learn More

3-Carene, Analytical Standard, ≥97.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018304
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018304 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018304 Supplier Merck Emd Millipore Supplier No. 944155ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 13466-78-9
Density 0.857 g/mL (at 25°C)
Physical Form Neat
Linear Formula C10H16
MDL Number MFCD00001315
Boiling Point 168°C to 169°C (705 mmHg)
Synonym delta3-Carene; 3,7,7-Trimethylbicyclo[4.1.0 ]hept-3-ene
Quantity 5 mL
Formula Weight 136.23
Percent Purity ≥97.0% (GC)
Grade Analytical Standard
Show More Show Less