missing translation for 'onlineSavingsMsg'
Learn More

3-Carboxyphenyl isothiocyanate, 97%

Catalog No. AAL1132206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1132206 5 g
AAL1132203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1132206 Supplier Thermo Scientific Chemicals Supplier No. L1132206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2131-63-7 | C8H5NO2S | 179.193 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2131-63-7
Molecular Formula C8H5NO2S
Molecular Weight (g/mol) 179.193
MDL Number MFCD00041367
InChI Key PJRBPKOOGLKPFB-UHFFFAOYSA-N
Synonym 3-carboxyphenyl isothiocyanate, 3-carboxyphenylisothiocyanate, benzoic acid, 3-isothiocyanato, benzoic acid,3-isothiocyanato, acmc-20an7x, m-carboxyphenyl isothiocyanate, 3-isothiocyanato-benzoic acid, #
PubChem CID 75046
IUPAC Name 3-isothiocyanatobenzoic acid
SMILES C1=CC(=CC(=C1)N=C=S)C(=O)O

Specifications

Melting Point ∽165°C (decomposition)
Odor Irritating
Quantity 5 g
UN Number UN2923
Beilstein 2209003
Sensitivity Moisture sensitive
Formula Weight 179.2
Percent Purity 97%
Chemical Name or Material 3-Carboxyphenyl isothiocyanate
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only