missing translation for 'onlineSavingsMsg'
Learn More

3-Carboxy-4-methyl-5-propyl-2-furanpropanoic acid, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

$616.58 - $1491.83

Specifications

CAS 86879-39-2
Linear Formula C12H16O5
MDL Number MFCD08436079
Synonym 3-Carboxy-4-methyl-5-propyl-2-furanpropionic acid; CMPF; Propylurofuranic acid
Type Analytical Standard
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111015714
View Documents
Merck Emd Millipore
9083310MG
10 mg
Each for $616.58
Only null left
 
111015715
View Documents
Merck Emd Millipore
9083350MG
50 mg
Each for $1,491.83
Only null left
 
Description

Description

Specifications

Specifications

86879-39-2
MFCD08436079
Analytical Standard
≥95.0% (HPLC)
C12H16O5
3-Carboxy-4-methyl-5-propyl-2-furanpropionic acid; CMPF; Propylurofuranic acid
240.25
Analytical Standard
Videos
SDS
Documents

Documents