missing translation for 'onlineSavingsMsg'
Learn More

3-Carboxy-4-methyl-5-propyl-2-furanpropanoic acid, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015715
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015715 50 mg
111015714 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015715 Supplier Merck Emd Millipore Supplier No. 9083350MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 86879-39-2
Linear Formula C12H16O5
MDL Number MFCD08436079
Synonym 3-Carboxy-4-methyl-5-propyl-2-furanpropionic acid; CMPF; Propylurofuranic acid
Type Analytical Standard
Quantity 50 mg
Formula Weight 240.25
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard