missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Carboxy-4-methyl-5-propyl-2-furanpropanoic acid, ≥95.0% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 86879-39-2 |
| Linear Formula | C12H16O5 |
| MDL Number | MFCD08436079 |
| Synonym | 3-Carboxy-4-methyl-5-propyl-2-furanpropionic acid; CMPF; Propylurofuranic acid |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 240.25 |
| Percent Purity | ≥95.0% (HPLC) |
| Grade | Analytical Standard |