missing translation for 'onlineSavingsMsg'
Learn More

3-Carboxy-4-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C29451G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C29451G Supplier TCI America Supplier No. C29451G
Only null left

Specifications

Chemical Name or Material 3-Carboxy-4-fluorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 872460-12-3
Melting Point 219°C
Color White
Molecular Formula C7H6BFO4
Quantity 1 g
MDL Number MFCD03095129
Synonym 3-carboxy-4-fluorophenylboronic acid, 3-carboxy-4-fluorobenzeneboronic acid, 5-dihydroxyboranyl-2-fluorobenzoic acid, 4-fluoro-3-carboxyphenylboronic acid, benzoic acid, 5-borono-2-fluoro, 3-carboxyl-4-fluorophenylboronic acid, 5-borono-2-fluoro-benzoic acid, akos brn-0540, pubchem16787, acmc-209qj6
InChI Key DGORTXQWDDXSIQ-UHFFFAOYSA-N
SMILES OB(O)C1=CC(C(O)=O)=C(F)C=C1
IUPAC Name 5-(dihydroxyboranyl)-2-fluorobenzoic acid
Molecular Weight (g/mol) 183.93
PubChem CID 2763242
Formula Weight 183.93
Physical Form Crystalline Powder
Show More Show Less