missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Carboxy-1-(4-sulfophenyl)-5-pyrazolone Sodium Salt 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 500 g |
| Formula Weight | 306.22 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Carboxy-1-(4-sulfophenyl)-5-pyrazolone Sodium Salt |
Safety and Handling
| EINECSNumber | (5)-3450 |
| TSCA | Yes |