missing translation for 'onlineSavingsMsg'
Learn More

3-Butynyl p-Toluenesulfonate 97.0+%, TCI America™

Catalog No. T20465G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T20465G Supplier TCI America Supplier No. T20465G

Chemical Identifiers

CAS 23418-85-1
Molecular Formula C11H12O3S
Molecular Weight (g/mol) 224.274
MDL Number MFCD00041687
InChI Key STOASOOVVADOKH-UHFFFAOYSA-N
Synonym 3-butynyl p-toluenesulfonate, but-3-yn-1-yl 4-methylbenzenesulfonate, 3-butynyl tosylate, 3-butynyl 4-toluenesulfonate, 3-1-butynyl-4-toluenesulphonate, 3-butynyl 4-methylbenzenesulfonate, p-toluenesulfonic acid 3-butynyl ester, but-3-ynyl tosylate, 4-tosyloxy-1-butyne, acmc-1cgfq
PubChem CID 272020
IUPAC Name but-3-ynyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCCC#C

Specifications

Quantity 5 g
Formula Weight 224.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Butynyl p-Toluenesulfonate