missing translation for 'onlineSavingsMsg'
Learn More

3-Butyn-1-ol, 97%

Catalog No. AC157331000
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC157331000 100 mL Glass bottle
AC157330050 5 mL Glass bottle
AC157330250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC157331000 Supplier Thermo Scientific Chemicals Supplier No. 157331000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 927-74-2 | C4H6O | 70.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 927-74-2
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00002955
InChI Key OTJZCIYGRUNXTP-UHFFFAOYSA-N
Synonym 3-butyn-1-ol, 3-butynol, 3-butyne-1-ol, 4-hydroxy-1-butyne, 3-butynyl alcohol, 1-butyn-4-ol, 2-hydroxyethylacetylene, homopropargyl alcohol, 2-hydroxyethyl acetylene, 4-hydroxy-butyne
PubChem CID 13566
ChEBI CHEBI:27444
IUPAC Name but-3-yn-1-ol
SMILES C#CCCO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -63.6°C
Color Colorless to Yellow
Density 0.9260g/mL
Boiling Point 128.8°C
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4397 to 1.4417
Linear Formula HC≡CCH2CH2OH
Quantity 100 mL
Beilstein 01, IV, 2219
Specific Gravity 0.926
Solubility Information Solubility in water: miscible
Formula Weight 70.09
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Butyn-1-ol, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-161-9
RTECSNumber ES0710000
TSCA TSCA

RUO – Research Use Only